Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD155/PVR Fc Chimera Protein

Catalog No. 9174CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-74C-D050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-74C-D050 Supplier R&D Systems™ Supplier No. 9174CD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human CD155/PVR Fc Chimera is immobilized at 1 μg/mL (100 μL/well), the concentration of Recombinant Human CD96 v2 Fc Chimera (Catalog # 9556-CD ) that produces 50% of the optimal binding response is 30-150 ng/mL.

Specifications

Accession Number NP_006496
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5817
Molecular Weight (g/mol) M.W. (Observed): 81-98 kDa, reducing conditions; M.W. (theoretical): 61 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human CD155/PVR protein Human CD155/PVR (Asp28-Asn343) (N-terminus) Accession # NP_006496 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4
Common Name CD155/PVR
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human CD155/PVR Fc Chimera is immobilized at 0.5μg/mL (100μL/well), the concentration of Recombinant Human DNAM-1/CD226 Fc Chimera (Catalog # 666-DN) that produces 50% of the optimal binding response is approximately 20-120ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Human CD155/PVR (Asp28-Asn343) Accession # NP_006496, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD155/PVR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.