Learn More
R&D Systems™ Recombinant Human CD177 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | AAH29167.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 57126 |
| Molecular Weight (g/mol) | M.W.-observed: 83-93 kDa, under reducing conditions., M.W.-theroretical: 68 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human CD177 protein Human CD177 (Leu22-Gly407) (C-terminus) Accession # AAH29167.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD177, CD177 antigenNB1 GP, CD177 molecule, HNA-2 A, HNA2Acell surface receptor, Human neutrophil alloantigen 2 A, NB1 GP, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1, PRV1NB1 glycoprotein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.