Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD19 Fc Chimera Alexa Fluor™ 488 Protein

Catalog No. AFG9269020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFG9269020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFG9269020 Supplier R&D Systems™ Supplier No. AFG9269020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by flow cytometry for its ability to bind anti-Human CD19 Monoclonal Antibody conjugated beads. The concentration of Recombinant Human CD19 Fc Chimera Alexa Fluor™ 488 (Catalog # AFG9269) that produces 50% of the binding is 0.10-1.50 μg/mL.

Specifications

Accession Number P15391.6
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 930
Molecular Weight (g/mol) M.W.-observed: 77-92 kDa, reducing conditions, M.W.-theroretical: 57 kDa
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human CD19 protein Human CD19 (Glu21-Lys291) (N-terminus) Accession # P15391.6 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12
Conjugate Alexa Fluor 488
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD19
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.