Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD2 His Protein
SDP

Catalog No. NBP222745 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222745 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222745 Supplier R&D Systems™ Supplier No. NBP222745

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-209 of Human CD2 The Recombinant Human CD2 Protein is derived from E. coli. The Recombinant Human CD2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001758
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 914
Molecular Weight (g/mol) 23.8kDa
Name CD2 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CD2, 25-209aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMKEITNA LETWGALGQD INLDIPSFQM SDDIDDIKWE KTSDKKKIAQ FRKEKETFKE KDTYKLFKNG TLKIKHLKTD DQDIYKVSIY DTKGKNVLEK IFDLKIQERV SKPKISWTCI NTTLTCEVMN GTDPELNLYQ DGKHLKLSQR VITHKWTTSL SAKFKCTAGN KVSKESSVEP VSCPEKGLD
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.