Learn More
R&D Systems™ Recombinant Human CD24 Fc Chimera Protein
Description
When Recombinant Human CD24 Fc Chimera is coated at 5 μg/mL, the concentration of Recombinant Human Siglec-10 Fc Chimera (Catalog # 2130-SL) that produces 50% optimal binding response is found to be approximately 0.3-3 μg/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 100133941 |
| Molecular Weight (g/mol) | 29.7 kDa (monomer) |
| Name | CD24 Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human CD24 protein, (N-terminus) Human CD24 (Met1-Gly59) Accession &Num; NP_037362 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.