Learn More
R&D Systems™ Recombinant Human CD2F-10/SLAMF9 Fc Chimera Protein
Description
Order Info
The product is shipped at ambient temperature, upon receipt, store immediately
Specifications
Specifications
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human CD2F-10/SLAMF9 protein, (N-terminus) Human CD2F-10/SLAMF9 (Arg19-Ser210) Accession &Num; Q96A28 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.