Learn More
R&D Systems™ Recombinant Human CD3 delta/CD3d Fc Avi-tag Protein
Description
Specifications
Specifications
| Accession Number | P04234.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 915 |
| Molecular Weight (g/mol) | M.W.-observed: 54-61 kDa, under reducing conditions, M.W.-theroretical: 38 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human CD3 delta/CD3d protein Human CD3 delta/CD3d (Phe22-Ala105) (c-terminus) Accession # P04234.1 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag N-terminus C-terminμs |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD3 antigen, delta subunit, CD3D, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.