Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD3 delta/CD3d Fc Chimera Protein

Catalog No. 10280TC050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-280-TC050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-280-TC050 Supplier R&D Systems™ Supplier No. 10280TC050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human CD3 delta/CD3d Fc Chimera is immobilized at 1.0 μg/mL, 100 μL/well, the concentration of Recombinant Human CD3E Fc Chimera Biotinylated Protein that produces 50% of the optimal binding response is approximately 1.0 to 6.0 μg/mL

Specifications

Accession Number P04234
For Use With (Application) Bioactivity
Gene ID (Entrez) 915
Molecular Weight (g/mol) 36 kDa
Name CD3 antigen, delta subunit, CD3D, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human CD3 delta/CD3d protein

Human CD3 delta/CD3d (Phe22-Ala105) Accession # P04234IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.