Learn More
Description
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. PBS (pH 7.4) containing 10% glycerol |
| Gene ID (Entrez) | 917 |
| Molecular Weight (g/mol) | 11.8 kDa |
| Name | Recombinant Human CD3 gamma Protein |
| Purification Method | >90%, by SDS-PAGE |
| Quantity | 10 μg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
