Learn More
R&D Systems™ Recombinant Human CD30/TNFRSF8 Fc Chimera Biotin Protein, CF
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 943 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 80-115 kDa, under reducing conditions., MolecularWeight-theroretical: 65 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD30/TNFRSF8 protein Human CD30/TNFRSF8 (Phe19-Lys379) (N-terminus) Accession # P28908.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 |
| Conjugate | Biotin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.