Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD300e Fc Chimera Alexa Fluor™ 647 Protein

Catalog No. AFR10479020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFR10479020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFR10479020 Supplier R&D Systems™ Supplier No. AFR10479020
Add to Cart
Add to Cart
This item is not returnable. View return policy

LMIR6

Measured by flow cytometry for its ability to bind anti-Human CD300e/LMIR6 Monoclonal Antibody conjugated beads.The concentration of Recombinant Human CD300e/LMIR6 Chimera Alexa Fluor™ 647 (Catalog # AFR10479) that produces 50% of the binding response is 0.20-4.00 ng/mL.

Specifications

Accession Number Q496F6.2
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 342510
Molecular Weight (g/mol) M.W.-observed: 45-58 kDa and 106-125 kDa (non-reducible dimer), under reducing conditions., M.W.-theroretical: 44 kDa
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived human CD300e/LMIR6 protein Human CD300e/LMIR6 (Leu18-Leu169) (N-terminus) Accession # Q496F6.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias CD300 antigen like family member E, CD300 antigen-like family member E, CD300e, CD300e antigenPIgR-2, CD300e molecule, CD300LE, CD300LEPIgR2, CLM2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM-2, IREM2IREM-2, LMIR6, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2
Conjugate Alexa Fluor 647
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD300e/LMIR6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.