Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD302/CLEC13A Fc Chimera Protein
SDP

Catalog No. 10203CL050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-203-CL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-203-CL050 Supplier R&D Systems™ Supplier No. 10203CL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

CD302, also known as CLEC13A and DCL1, is a type I transmembrane C-type lectin receptor

  • Identified while cloning human DEC-205 and was termed DCL1 (DEC-205-associated C-type lectin-1)
  • In humans, the highest expression of CD302 transcripts was observed in the liver, followed by lungs, spleen, and myeloid PBMC populations including monocytes, granulocytes, and dendritic cells

Specifications

Accession Number Q8IX05-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 9936
Molecular Weight (g/mol) 43 kDa
Purification Method >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human CD302/CLEC13A protein

Human CD302/CLEC13A (Asp23-Asn167) Accession # Q8IX05-1IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.