Learn More
R&D Systems™ Recombinant Human CD38 His-tag Avi-tag Protein
Description
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 952 |
| Molecular Weight (g/mol) | M.W.-observed: 40-52 kDa, under reducing conditions, M.W.-theoretical: 33 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD38 protein Human CD38 (Val43-Ile300) (N-terminus) Accession # P28907.2 HHHHHH Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | 2 '-phospho-ADP-ribosyl cyclase, 2 '-phospho-ADP-ribosyl cyclase/2 '-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2 '-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl Cyclase, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose Hydrolase, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 |
| Conjugate | Biotin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.