Learn More
R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (HEK293-expressed)
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | P29965.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS and EDTA with Trehalose. |
| Gene ID (Entrez) | 959 |
| Molecular Weight (g/mol) | M.W.-observed: 20-27 kDa, under reducing conditions., M.W.-theroretical: 22 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD40 Ligand/TNFSF5 protein YPYDVPDYA GCN4-IZ GGGSGGGSGGGS Human CD40 Ligand (Met113-Leu261) (C-terminus) Accession # P29965.1 (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40L, CD40-L, CD40LG, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5, TNFSF5IMD3, TRAP, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.