Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD47 Fc Chimera Protein
SDP

Catalog No. 4670CD050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4670CD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4670CD050 Supplier R&D Systems™ Supplier No. 4670CD050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human (rh) SIRP alpha/CD172a Fc Chimera (Catalog # 4546-SA ) is coated at 2 μg/mL, Recombinant Human CD47 Fc Chimera binds with an apparent Kd <1 nM.

Optimal dilutions should be determined by each laboratory for each application.

Also measured by its ability to antagonize red blood cell adhesion to immobilized rhSIRP alpha Fc Chimera. The ED50 for this effect is 0.5-2.0 μg/mL.

Specifications

Accession Number Q08722
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 961
Molecular Weight (g/mol) M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 40.3 kDa (monomer)
Name CD47 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human CD47 protein Human CD47 (Gln19-Pro139) (N-terminus) Accession # Q08722 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD47
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD47
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.