Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD48/SLAMF2 Fc Chimera Protein
SDP

Catalog No. 9310CD050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-10C-D050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-10C-D050 Supplier R&D Systems™ Supplier No. 9310CD050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CD48/SLAMF2 Fc Chimera Protein is derived from HEK293. The Recombinant Human CD48/SLAMF2 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature

Specifications

Accession Number P09326
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 962
Molecular Weight (g/mol) 49 kDa
Name CD48/SLAMF2 Fc Chimera Recombinant Protein
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human CD48/SLAMF2 protein, (N-terminus) Human CD48/SLAMF2 (Gln27-Ser220) Accession &Num; P09326 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Common Name BCM1 surface antigen; BCM1; BCM1Leukocyte antigen MEM-102; BLAST; BLAST1; BLAST1TCT.1; B-lymphocyte activation marker BLAST-1; CD48 antigen (B-cell membrane protein); CD48 antigen; CD48 molecule; CD48; hCD48; mCD48; MEM-102; SLAMF2
Biological Activity 2 to 12ng/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.