Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD68/SR-D1 His Protein
SDP

Catalog No. NBP222756 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222756 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222756 Supplier Novus Biologicals™ Supplier No. NBP222756
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 22-319 of Human CD68/SR-D1 The Recombinant Human CD68/SR-D1 Protein is derived from E. coli. The Recombinant Human CD68/SR-D1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001242
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 1 M UREA, 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 968
Molecular Weight (g/mol) 34.1kDa
Name CD68/SR-D1 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen CD68, 22-319aa, Human, His tag, E.coli MGSSHPLHHH SSGLVPRGSH MGSHMNDCPH KKSATLLPSF TVTPTVTEST GTTSHRTTKS HKTTTHRTTT TGTTSHGPTT ATHNPTTTSH GNVTVHPTSN STATSQGPST ATHSPATTSH GNATVHPTSN STATSPGFTS SAHPEPPPPS PSPSPTSKET IGDYTWTNGS QPCVHLQAQI QIRVMYTTQG GGEAWGISVL NPNKTKVQGS CEGAHPHLLL SFPYGHLSFG FMQDLQQKVV YLSYMAVEYN VSFPHAAQWT FSAQNASLRD LQAPLGQSFS CSNSSIILSP AVHLDLLSLR LQAAQLPHTG VFGQSFSCPS DRS
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Cell Biology, Cytokine Research, Immunology, Microglia Markers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.