Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD69 Protein

Catalog No. 8468CD025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-68C-D025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-68C-D025 Supplier R&D Systems™ Supplier No. 8468CD025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CD69 Protein is derived from NS0. The Recombinant Human CD69 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q07108
For Use With (Application) Bioactivity
Gene ID (Entrez) 969
Molecular Weight (g/mol) Observed M.W.: 20-30kDa, reducing conditions, Theoretical M.W.: 17kDa
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human CD69 protein Gly64-Lys199 with an N-terminal 9-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, EA-1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leu23, Leukocyte surface antigen Leu-23, MLR-3, p60
Common Name CD69
Gene Symbol CD69
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human CD69 is coated at 5μg/mL (100μL/well), the concentration of Recombinant Human Galectin-1 that produces a 50% optimal binding response is typically 0.4-2μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Gly64-Lys199 with an N-terminal 9-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.