Learn More
Creative Biomart Recombinant Human CD70 protein, His-tag, GMP Grade
Supplier:  Creative Biomart CD70328THP
Encompass_Preferred
Recombinant Human CD70 protein GMP Grade(NP_001243)(154 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5320
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.