Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD8 alpha/beta Heterodimer Protein

Catalog No. 9358CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-58C-D050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-58C-D050 Supplier R&D Systems™ Supplier No. 9358CD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CD8 alpha/beta Heterodimer Protein is derived from HEK293. The Recombinant Human CD8 alpha/beta Heterodimer Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P01732 (CD8 alpha), P10966 (CD8 beta)
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 925
Molecular Weight (g/mol) 26 kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human CD8 protein, (N-terminus) Human CD8 alpha (Ser22-Asp182) Accession &Num; P01732 Acidic Tail HHHHHH Human CD8 beta (Asn19-Pro170) Accession &Num; P10966 Basic Tail FLAG (DYKDDDDK) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked heterodimer
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Biological Activity 0.8 to 4μg/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.