Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD8 beta His Protein
SDP

Catalog No. p-7104435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25153605 0.5 mg
NBP25153601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25153605 Supplier Novus Biologicals™ Supplier No. NBP2515360.5MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 22-170 of Human CD8 beta The Recombinant Human CD8 beta Protein is derived from E. coli. The Recombinant Human CD8 beta Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.4M UREA, 10% glycerol
Gene ID (Entrez) 926
Molecular Weight (g/mol) TMW: 19.2kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Common Name CD8 beta
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.