Learn More
R&D Systems™ Recombinant Human CD94 Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | Q13241 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 26253 |
| Molecular Weight (g/mol) | 18 kDa, reducing conditions |
| Name | DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68, M68DJ583P15.1.1, TNFRSF6B, TR6, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human CD94 protein Lys32-Ile179,with an N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.