Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CDK4 His Protein
SDP

Catalog No. NBP222974 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222974 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222974 Supplier R&D Systems™ Supplier No. NBP222974

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-303 of Human CDK4 The Recombinant Human CDK4 Protein is derived from E. coli. The Recombinant Human CDK4 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000066
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 1019
Molecular Weight (g/mol) 36.1kDa
Name CDK4 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CDK4, 1-303aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMATSRYE PVAEIGVGAY GTVYKARDPH SGHFVALKSV RVPNGGGGGG GLPISTVREV ALLRRLEAFE HPNVVRLMDV CATSRTDREI KVTLVFEHVD QDLRTYLDKA PPPGLPAETI KDLMRQFLRG LDFLHANCIV HRDLKPENIL VTSGGTVKLA DFGLARIYSY QMALTPVVVT LWYRAPEVLL QSTYATPVDM WSVGCIFAEM FRRKPLFCGN SEADQLGKIF DLIGLPPEDD WPRDVSLPRG AFPPRGPRPV QSVVPEMEES GAQLLLEMLT FNPHKRISAF RALQHSYLHK DEGNPE
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.