Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Cdk5 His Protein
SDP

Catalog No. NB398304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398304 0.01 mg
NB398303 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB398304 Supplier Novus Biologicals™ Supplier No. NBP2611690.010MG
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 40% glycerol, 1 mM DTT
Gene ID (Entrez) 1020
Molecular Weight (g/mol) MolecularWeight-theroretical: 34.1 kDa
Quantity 0.01 mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less Than 1.0 EU / 1 μg of protein (determined by LAL method)
Gene Alias Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit
Gene Symbol CDK5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Form Purified
Research Category Innate Immunity
Purity or Quality Grade >90%, by SDS-PAGE
Protein Cdk5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.