Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CEACAM-5/CD66e Protein
SDP

Catalog No. 4128CM050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4128CM050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4128CM050 Supplier R&D Systems™ Supplier No. 4128CM050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag

Measured by its binding activity in a functional ELISA. Ideo, H. et al. (2005) J. Biol. Chem. 280:4730. Recombinant Human CEACAM-5/CD66e can bind Recombinant Human Galectin-4 (Catalog # 1227-GA with an estimated Kd <4 nM.

Specifications

Accession Number Q8N4D0
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1048
Molecular Weight (g/mol) M.W. (Observed): 155-180 kDa, reducing conditions; M.W. (theoretical): 72 kDa
Name CEACAM-5/CD66e
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human CEACAM-5/CD66e protein Lys35-Ala685, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol CEACAM5
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CEACAM5/CD66e
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.