Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CEACAM-7 Protein

Catalog No. 9010CM050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-10C-M050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-10C-M050 Supplier R&D Systems™ Supplier No. 9010CM050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CEACAM-7 Protein is derived from CHO. The Recombinant Human CEACAM-7 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q14002
For Use With (Application) Bioactivity
Gene ID (Entrez) 1087
Molecular Weight (g/mol) Observed M.W.: 42-52kDa, reducing conditions, Theoretical M.W.: 23kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human CEACAM-7 protein Thr36-Asn233,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CEACAM7, CGM 2, CGM2
Common Name CEACAM-7
Gene Symbol CEACAM7
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human CEACAM-7 is immobilized at 0.1μg/mL (100μL/well), the concentration of Recombinant Human CEACAM-1/CD66a that produces 50% of the optimal binding response is approximately 1.6-8ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Thr36-Asn233, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.