Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Chitinase 3-like 2 Protein

Catalog No. 5112CH050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5112CH050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5112CH050 Supplier R&D Systems™ Supplier No. 5112CH050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by the ability of the immobilized protein to support the adhesion of FaDu human squamous cell carcinoma cells.

When 5 x 104 cells per well are added to rhCHI3L2 coated plates for 30 minutes at 37 °C, cell adhesion is enhanced in a dose dependent manner with typical ED50 range of 0.7-2.8 μg/mL.

Opitimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number Q15782
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1117
Molecular Weight (g/mol) M.W. (Observed): 40 kDa, reducing conditions; M.W. (theoretical): 42.2 kDa
Name Chitinase 3-like 2
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Chitinase 3-like 2 protein Tyr27-Leu390, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol CHI3L2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Chitinase 3-like 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.