Learn More
R&D Systems™ Recombinant Human cIAP-2 (HIAP-1) Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 330 |
| Molecular Weight (g/mol) | 71 kDa |
| Name | cIAP-2/HIAP-1 |
| Quantity | 50 μg |
| Source | E. coli-derived human cIAP-2/HIAP-1 protein, (N-terminus) ATVID 10-His tag SIEGRA Human cIAP-2 (Asn2-Ser604) Accession &Num; Q13489 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.