Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CLEC2L Fc Chimera Protein
SDP

Catalog No. 10204CL050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-204-CL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-204-CL050 Supplier R&D Systems™ Supplier No. 10204CL050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

C-type lectin domain family 2, member L (CLEC2L), is a type II transmembrane protein of the CLEC2 family of proteins that includes CD69, LLT1, AICL and KACL

  • CLEC2 family members have broad tissue expression, with LLT1, AICL and KACL expressed on B cells, monocytes, and keratinocytes, respectively
  • In contrast, CLEC2L, a homodimeric cell surface receptor, is predominantly expressed in the brain and is referred to as the brain-associated C-type lectin (BACL)
  • Mature human CLEC2L includes a 68 amino acid (aa) cytoplasmic domain, a 21 aa transmembrane segment, and a 125 aa extracellular domain (ECD).
  • Within the ECD, human CLEC2L shares 94% aa sequence identity with mouse and rat CLEC2L.

Specifications

Accession Number P0C7M8-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 154790
Molecular Weight (g/mol) 41 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human CLEC2L protein

MDHuman IgG2A (Pro100-Lys333)IEGRHuman CLEC2L (Ser90-Thr214) Accession # P0C7M8-1N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.