Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS. |
| Gene ID (Entrez) | 1270 |
| Molecular Weight (g/mol) | 22.8 kDa |
| Name | CNTF |
| Quantity | 10 μg |
| Source | E. coli-derived human CNTF protein Ala2-Met200 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. |
| Structural Form | This recombinant protein is prone to proteolytic degradation at the C-terminus, resulting in proteins with molecular masses of 22.2 - 22.8 kDa. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
