Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CNTFR/CNTF Complex Protein

Catalog No. 10283CC020
Change view
Click to view available options
Quantity:
20 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-283-CC020 20 μg
10-283-CC050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-283-CC020 Supplier R&D Systems™ Supplier No. 10283CC020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Up to 100-fold greater activity than CNTF alone

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells.

Specifications

Accession Number P26992 & P26441
For Use With (Application) Bioactivity
Molecular Weight (g/mol) 59 kDa
Quantity 20 μg
Source

Human embryonic kidney cell, HEK293-derived human CNTFR/CNTF Complex protein

Human CNTFR (Gln23-Ser342) Accession # P26992GGGSGGGSGGGSHuman CNTF (Met1-Met200) Accession # P26441N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.