Learn More
R&D Systems™ Recombinant Human CNTFR/CNTF Complex Protein
Description
Specifications
Specifications
| Accession Number | P26992 & P26441 |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | 59 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CNTFR/CNTF Complex protein Human CNTFR (Gln23-Ser342) Accession # P26992GGGSGGGSGGGSHuman CNTF (Met1-Met200) Accession # P26441N-terminus C-terminus |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Species | Human |
| Reconstitution | Reconstitute at 200 μg/mL in PBS. |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.