Learn More
R&D Systems™ Recombinant Human Coagulation Factor II/Thrombin Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in HEPES and NaCl. |
| Gene ID (Entrez) | 2147 |
| Molecular Weight (g/mol) | 4 kDa (light chain A), 31 kDa (heavy chain B), 14 kDa (C-terminal fragment) |
| Name | Coagulation Factor II/Thrombin |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Coagulation Factor II/Thrombin protein Met1-Glu622 with a C-terminal 10-His tag The proenzyme was purified,activated and further purified. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Active form |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.