Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Coagulation Factor Xa Protein
SDP

Catalog No. 1063SE010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1063SE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1063SE010 Supplier R&D Systems™ Supplier No. 1063SE010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Coagulation Factor Xa Protein is derived from Sf 21 (baculovirus). The Recombinant Human Coagulation Factor Xa Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MES, NaCl and CaCl 2 .
Gene ID (Entrez) 2159
Molecular Weight (g/mol) 12 kDa & 30 kDa
Name Coagulation Factor X
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Coagulation Factor X protein Leu24-Lys488 with a C-terminal 10-His tag Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Active form
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 25 mM MES, 150 mM NaCl and 5 mM CaCl 2 pH 6.0.
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.