Learn More
R&D Systems™ Recombinant Human Collagen XV alpha 1/COL15A1 Protein
Shop All Novus Biologicals Products

Description
- Derived from HEK293.
- Validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P39059 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 1306 |
| Molecular Weight (g/mol) | 26 to 36 kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Collagen XV alpha 1/COL15A1 protein. Hemagglutinin Tag (YPYDVPDYA)Human COL15A1 (Asn1133-Ser1381) Accession # P39059 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.