Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Common gamma Chain Protein

Catalog No. 384RG050
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
384RG050 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
384RG050CF Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 384RG050 Supplier R&D Systems™ Supplier No. 384RG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Common gamma Chain Protein is derived from Sf 21 (baculovirus). The Recombinant Human Common gamma Chain Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3561
Molecular Weight (g/mol) 28.2 kDa
Name Common gamma Chain/IL-2 R gamma
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Common gamma Chain/IL-2 R gamma protein Leu23-Asn254,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Activity Measured by its ability to inhibit the IL-2-dependent proliferation of MO7e human megakaryocytic leukemic cells. The ED50 for this effect is typically 0.6-3.6 μg/mL in the presence of 30 μg/mL of soluble IL-2 R beta and 10 ng/mL of recombinant human IL-2.
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.