Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CtBP1 Protein
SDP

Catalog No. NBP141161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141161 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141161 Supplier Novus Biologicals™ Supplier No. NBP141161
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-440 of Human CtBP1 The Recombinant Human CtBP1 Protein is derived from E. coli. The Recombinant Human CtBP1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q13363
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 100mM NaCl, 10% glycerol
Gene ID (Entrez) 1487
Molecular Weight (g/mol) 47.5kDa
Name CtBP1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen CtBP1, 1-440aa, Human, E.coli (NP_001319).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.