Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CTRP3/C1qTNF3/CORS26 Protein

Catalog No. 9398TN025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
9398TN025 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
9398TN025CF Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 9398TN025 Supplier R&D Systems™ Supplier No. 9398TN025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

  • Measured by its ability to induce phospho AMPK activation in C2C12 mouse differentiated myocytes.
  • 300ng/mL of Recombinant Human CTRP3/C1qTNF3/CORS26 induces phosphorylation of AMPK.

Specifications

Accession Number Q9BXJ4-1
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 114899
Molecular Weight (g/mol) 24kDa
Quantity 25 μg
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias C1ATNF3, C1q and tumor necrosis factor related protein 3, C1qTNF3, Cartducin, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.