Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CTRP9/C1qTNF9 Protein
SDP

Catalog No. 6537TN050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-37T-N050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 65-37T-N050 Supplier R&D Systems™ Supplier No. 6537TN050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human CTRP9/C1qTNF9 Protein is derived from NS0. The Recombinant Human CTRP9/C1qTNF9 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAH40438
Formulation Lyophilized from a 0.2μm filtered solution in HEPES and NaCl
Gene ID (Entrez) 338872
Molecular Weight (g/mol) 34kDa
Name C1qTNF9
Preparation Method Reconstitute at 250μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 36-40kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human CTRP9/C1qTNF9 protein Asn16-Pro333,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination His
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.