Learn More
Description
Specifications
Specifications
| Accession Number | Q13618, Q15843, P62877 |
| For Use With (Application) | Bioactivity |
| Formulation | X mg/ml (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL3/NEDD8/RBX1 protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 60 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Protein | CUL3/NEDD8/RBX1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
