Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CUL4A/RBX1 Complex Protein

Catalog No. E3440025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3440025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3440025 Supplier R&D Systems™ Supplier No. E3440025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Typical enzyme concentration to support in vitro conjugation will depend on experimental conditions.

Specifications

Accession Number Q13619.3 (CUL4A), P62877.1 (RBX1)
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, Glycerol and DTT.
Molecular Weight (g/mol) M.W. Theroretical: 94 kDa (CUL4A), 12 kDa (RB x 1)
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL4A/RBX1 Complex protein Met1 - Ala759 with a N-terminal 10-His Sumo tag (CUL4A), Met1 - His108 (RBX1)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity CUL4A/RBX1 Complex
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer X mg/mL (X μm) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein CUL4A/RBX1 Complex
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.