Learn More
Boston Biochem™ Recombinant Human CUL5/RBX2 Complex Protein, CF
Description
Specifications
Specifications
| Accession Number | Q93034, Q9UBF6 |
| For Use With (Application) | Bioactivity |
| Formulation | X mg/mL (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT |
| Molecular Weight (g/mol) | M.W. (theoretical): 94 kDa (CUL5), 13 kDa (RB x 2) |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL5/RBX2 Complex protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 60 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
| Protein | CUL5/RBX2 Complex |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.