Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CXCL1/GRO alpha (aa 39-107) Protein
SDP

Catalog No. 9196GR025CF Shop All R&D Systems Products
Change view
Click to view available options
Formulation:
With BSA as carrier
Without BSA as carrier
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
91-96G-R025CF Without BSA as carrier
91-96G-R025 With BSA as carrier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 91-96G-R025CF Supplier R&D Systems™ Supplier No. 9196GR025/CF
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CXCL1/GRO alpha (aa 39-107) Protein is derived from E. coli. The Recombinant Human CXCL1/GRO alpha (aa 39-107) Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

The product is shipped at ambient temperature.

Specifications

Accession Number Q5M8T4
For Use With (Application) Bioactivity
Formulation Without BSA as carrier
Gene ID (Entrez) 1490
Molecular Weight (g/mol) 6 kDa, reducing conditions
Quantity 25 μg
Source E. coli-derived human CXCL1/GRO alpha/KC/CINC-1 protein Thr39-Asn107
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Biological Activity 0.15 to 0.9ng/mL (ED 50)
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag No
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.