Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CXCL3/GRO gamma (aa 39-107) Protein

Catalog No. 8297GG025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
82-97G-G025 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
82-97G-G025CF Lyophilized from a 0.2μm filtered solution in PBS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 82-97G-G025 Supplier R&D Systems™ Supplier No. 8297GG025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CXCL3/GRO gamma (aa 39-107) Protein is derived from E. coli. The Recombinant Human CXCL3/GRO gamma (aa 39-107) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P19876
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Gene ID (Entrez) 2921
Molecular Weight (g/mol) 8kDa
Name CXCL3/GRO gamma/CINC-2/DCIP-1 (aa 39-107)
Preparation Method Reconstitute at 250μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 7kDa, reducing conditions
Quantity 25 μg
Source E. coli-derived human CXCL3/GRO gamma/CINC-2/DCIP-1 protein Thr39-Asn107
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Thr39
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.