Learn More
Novus Biologicals™ Recombinant Human CXCL3/GRO gamma/CINC-2/DCIP-1 Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | P19876, P19876, P19876 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 50 mM NaCl. |
| Gene ID (Entrez) | 2921 |
| Molecular Weight (g/mol) | 7.9 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CXCL3/GRO gamma/CINC-2/DCIP-1 as determined by LAL method. |
| Gene Alias | chemokine (C-X-C motif) ligand 3, CINC-2 alpha, CINC-2 alpha/CXCL3, CINC-2 beta, CINC-2 beta/CXCL3, CINC-2b, C-X-C motif chemokine 3, DCIP-1, DCIP-1/CXCL3, GRO3, GRO3 oncogene, GROg, GRO-gamma, GRO-gamma(1-73), Growth-regulated protein gamma, Macrophage inflammatory protein 2-beta, melanoma growth stimulatory activity gamma, MGSA gamma, MIP-2b, MIP2B, MIP2-beta, SCYB3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.