Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CXCL5/ENA-78 Protein

Catalog No. 254XB025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS.
Lyophilized from a 0.2mm filtered solution in PBS containing BSA as carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
254XB025 Lyophilized from a 0.2mm filtered solution in PBS containing BSA as carrier protein
254XB025CF Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 254XB025 Supplier R&D Systems™ Supplier No. 254XB025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human CXCL5/ENA-78 Protein is derived from E. coli. The Recombinant Human CXCL5/ENA-78 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2mm filtered solution in PBS containing BSA as carrier protein
Gene ID (Entrez) 6374
Molecular Weight (g/mol) 8.4 kDa
Name CXCL5/ENA-78
Quantity 25 μg
Source E. coli-derived human CXCL5/ENA-78 protein Ala37-Asn114
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Activity Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CXCR2. The ED50 is 3-15 ng/mL
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.