Learn More
Eurofins Discoverx Recombinant Human CXCL8, 5 µg. (92-1009)
Supplier:  Eurofins Discoverx 921009
Encompass_Preferred
CXCL8, aka IL-8, is a cytokine protein produced by macrophages and other cells such as epithelial cells, endothelial cells, and airway myocytes. IL-8 is able to induce chemotaxis in neutrophils and granulocytes, attracting them to the site of damage or infection. IL-8 can also induce phagocytosis of foreign debris, and is known to be a potent promoter of angiogenesis. It is an agonist for CXCR1 and CXCR2, with more affinity for the former. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. NC1464904
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.