Learn More
Description
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
| Gene ID (Entrez) | 27074 |
| Molecular Weight (g/mol) | TMW: 38.8kDa |
| Quantity | 0.05 mg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 |
| Common Name | DC-LAMP |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
