Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human DDB1 Protein, CF

Catalog No. E3501050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3501050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3501050 Supplier R&D Systems™ Supplier No. E3501050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results

Specifications

Accession Number Q16531.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 1642
Molecular Weight (g/mol) M.W. (theoretical): 127 kDa
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human DDB1 protein Met1 - His1140
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein DDB1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.