Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human DDX39 His Protein
SDP

Catalog No. NBP25173105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25173105 0.5 mg
NBP25173101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25173105 Supplier Novus Biologicals™ Supplier No. NBP2517310.5MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-249 of Human DDX39 The Recombinant Human DDX39 Protein is derived from E. coli. The Recombinant Human DDX39 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 0.4M UREA
Gene ID (Entrez) 10212
Molecular Weight (g/mol) TMW: 31kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias ATP-dependent RNA helicase DDX39, BAT1, BAT1L, DDX39, DDXL, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39 transcript, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A, DEAD box protein 39, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39, EC 3.6.4.13, MGC18203, MGC8417, Nuclear RNA helicase URH49, nuclear RNA helicase, DECD variant of DEAD box family, UAP56-related helicase, 49 kDa, URH49
Common Name DDX39
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.